Invitrogen
Manufacturer Code:PA143067
Catalog # PIPA143067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rat Factor IX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Christmas factor; coagulation factor 9; Coagulation factor IX; Coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; F9 Coagulation factor 9 FIX; factor 9; factor IX (Christmas factor) precursor; factor IX F9; plasma thromboplastic component; Plasma thromboplastin component; PTC
Gene Aliases: AW111646; Cf-9; Cf9; F9; F9 p22; FIX; HEMB; P19; PTC; THPH8
UniProt ID: (Rat) P16296, (Human) P00740, (Mouse) P16294
Entrez Gene ID: (Rat) 24946, (Bovine) 280688, (Human) 2158, (Mouse) 14071
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.