Invitrogen
Manufacturer Code:PA579035
Catalog # PIPA579035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat Factor D recombit protein (Position: I26-A263). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa adipocyte protein; Adipsin; C3 convertase activator; Complement factor D; complement factor D (adipsin); Complement factor D Adipsin C3 convertase activator Endogenous vascular elastase Properdin factor D Cfd Adn Df; complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Endogenous vascular elastase; Properdin factor D
Gene Aliases: ADIPSIN; ADN; CFD; DF; EVE; PFD
UniProt ID: (Human) P00746, (Rat) P32038, (Mouse) P03953
Entrez Gene ID: (Human) 1675, (Rat) 54249, (Mouse) 11537
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.