Abnova
Manufacturer Code:PAB19609
Catalog # 89-121-136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human FZR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of FZR1.
For Research Use Only
Protein Aliases: CDC20 homolog 1; CDC20-like 1b; CDC20-like protein 1; CDC20C; CDH1; Cdh1/Hct1 homolog; Fizzy-related protein homolog; FZR; FZR2; HCDH; HCDH1; KIAA1242
Gene Aliases: CDC20C; CDH1; FYR; FZR; FZR1; FZR2; HCDH; HCDH1; KIAA1242
UniProt ID: (Human) Q9UM11
Entrez Gene ID: (Human) 51343
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.