Proteintech
Manufacturer Code:163681AP150UL
Catalog # 50-173-0896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FZR1 Fusion Protein Ag9201 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC20 homolog 1; CDC20 like protein 1 CDC20C CDH1 Cdh1/Hct1 homolog Fizzy related protein homolog FYR FZR FZR1 FZR2 HCDH HCDH1 KIAA1242; CDC20-like 1b; CDC20-like protein 1; Cdh1/Hct1 homolog; cell division cycle 20 related 1; Fizzy-related protein homolog; Fzr; hCDH1
Gene Aliases: AW108046; CDC20C; CDH1; FYR; FZR; FZR1; FZR2; HCDH; HCDH1; KIAA1242
UniProt ID: (Human) Q9UM11, (Mouse) Q9R1K5
Entrez Gene ID: (Human) 51343, (Mouse) 56371, (Rat) 314642
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.