Invitrogen
Manufacturer Code:PA547151
Catalog # PIPA547151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Frizzled-6 His19-Val153 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled 6; Frizzled-6; Fz-6; FZD6 Frizzled family receptor 6 FZ-6 HFZ6 Frizzled 6 Frizzled-6 Frizzled homolog 6 FZ6 NDNC10; mFz6
Gene Aliases: Fz6; Fzd6
UniProt ID: (Mouse) Q61089
Entrez Gene ID: (Mouse) 14368
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.