Invitrogen
Manufacturer Code:PA533545
Catalog # PIPA533545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from N-terminal extracellular domain of human Frizzled-6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled 6, seven transmembrane spanning receptor; frizzled family receptor 6; frizzled homolog 6; Frizzled-6; Fz-6; FZD6 Frizzled family receptor 6 FZ-6 HFZ6 Frizzled 6 Frizzled-6 Frizzled homolog 6 FZ6 NDNC10; seven transmembrane helix receptor
Gene Aliases: FZ-6; FZ6; FZD6; HFZ6; NDNC10
UniProt ID: (Human) O60353
Entrez Gene ID: (Human) 8323
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.