Invitrogen
Manufacturer Code:PA533557
Catalog # PIPA533557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from N-terminal extracellular domain of human FZD5 / Frizzled 5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled 5, seven transmembrane spanning receptor; frizzled family receptor 5; Frizzled-5; Fz-5; FZD5 C2orf31 DKFZP434E2135 Frizzled family receptor 5 Fz5 Fzd-5 FzE5 Frizzled 5 Frizzled-5 Wnt receptor Fz-5 HFZ5; FzE5; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Aliases: C2orf31; FZD5; HFZ5
UniProt ID: (Human) Q13467
Entrez Gene ID: (Human) 7855
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.