Proteintech
Manufacturer Code:215191AP150UL
Catalog # 50-173-3460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FZD5 Fusion Protein Ag16082 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2orf31 Frizzled 5 Fz 5 FZD5 FzE5 HFZ5; frizzled 5, seven transmembrane spanning receptor; frizzled family receptor 5; Frizzled-5; Fz-5; FzE5; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Aliases: C2orf31; FZD5; HFZ5
UniProt ID: (Human) Q13467
Entrez Gene ID: (Human) 7855
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.