Invitrogen
Manufacturer Code:PA547228
Catalog # PIPA547228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Frizzled-4 Phe37-Glu180 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD344; Frizzled-4; Fz-4; FZD4 CD344 CD344 antigen EVR1 Frizzled family receptor 4 Frizzled-4 Fz-4 Fz4 HFz4 Exudative vitreoretinopathy 1 FEVR FZD4S FzE4 Frizzled 4 Frizzled homolog 4 WNT receptor frizzled-4; mFz4
Gene Aliases: Fz4; Fzd4
UniProt ID: (Mouse) Q61088
Entrez Gene ID: (Mouse) 14366
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.