Invitrogen
Manufacturer Code:PA541972
Catalog # PIPA541972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FZD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD344; frizzled 4, seven transmembrane spanning receptor; frizzled family receptor 4; Frizzled family receptor 4 EVR1 FEVR FZD4S Fz-4 FzE4 GPCR MGC34390 CD344; frizzled homolog 4; frizzled receptor 4; Frizzled-4; Fz-4; Fzd4; FzE4; mFz4; rFz4; WNT receptor frizzled-4
Gene Aliases: CD344; EVR1; FEVR; Fz-4; Fz4; FZD4; FZD4S; FzE4; GPCR; hFz4
UniProt ID: (Human) Q9ULV1, (Mouse) Q61088, (Rat) Q9QZH0
Entrez Gene ID: (Human) 8322, (Bovine) 445416, (Horse) 100050465, (Mouse) 14366, (Guinea pig) 100714941, (Rat) 64558
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.