Invitrogen
Manufacturer Code:PA533551
Catalog # PIPA533551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from N-terminal extracellular domain of human FZD10 / Frizzled 10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD350; frizzled 10, seven transmembrane spanning receptor; frizzled family receptor 10; frizzled homolog 10; Frizzled-10; Fz-10; FZD10 CD350 antigen CD350 FZ-10 Frizzled homolog 10 HFz10 Frizzled 10 Frizzled family receptor 10 Frizzled-10 FzE7 Fz10; FzE7
Gene Aliases: CD350; FZ-10; Fz10; FZD10; FzE7; hFz10
UniProt ID: (Human) Q9ULW2
Entrez Gene ID: (Human) 11211
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.