Invitrogen
Manufacturer Code:PA523159
Catalog # PIPA523159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (170 RPQRPHSAQEHPLKDG 185) of Frizzled-10 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD350; CD350 FZ-10 Fz10 FzE7 hFz10 frizzled 10 seven transmembrane spanning receptor frizzled homolog 10 frizzled-10; frizzled 10, seven transmembrane spanning receptor; frizzled family receptor 10; frizzled homolog 10; frizzled homolog 10, pseudogene 1; Frizzled-10; Fz-10; FzE7
Gene Aliases: CD350; FZ-10; Fz10; FZD10; Fzd10-ps1; FzE7; hFz10
UniProt ID: (Human) Q9ULW2, (Mouse) Q8BKG4
Entrez Gene ID: (Human) 11211, (Rat) 363913, (Mouse) 93897
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.