Abnova
Manufacturer Code:PAB25006
Catalog # 89-126-683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to FZD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of FZD1.
For Research Use Only
Protein Aliases: DKFZp564G072; frizzled 1, seven transmembrane spanning receptor; frizzled family receptor 1; frizzled homolog 1; frizzled, Drosophila, homolog of, 1; Frizzled-1; Fz-1; FzE1; hFz1; Wnt receptor
Gene Aliases: FZD1
UniProt ID: (Human) Q9UP38
Entrez Gene ID: (Human) 8321
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.