Novus Biologicals
Manufacturer Code:NBP204090
Catalog # NBP204090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human UAP56-Interacting Factor using the numbering given in entry NP_115664.2 (GeneID 84248). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: forty-two-three domain containing 1 Forty-two-three domain-containing protein 1 Protein 40-2-3 UAP56-interacting factor UIFDKFZp761B1514; Forty-two-three domain-containing protein 1; Protein 40-2-3; UAP56-interacting factor
Gene Aliases: FYTTD1; UIF
UniProt ID: (Human) Q96QD9
Entrez Gene ID: (Human) 84248
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.