Invitrogen
Manufacturer Code:PA541057
Catalog # PIPA541057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FXYD7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FXYD domain containing ion transport regulator 7 FLJ25096; FXYD domain-containing ion transport regulator 7
Gene Aliases: FXYD7
UniProt ID: (Human) P58549, (Bovine) Q3ZBJ3
Entrez Gene ID: (Human) 53822, (Bovine) 616128
Molecular Function: enzyme modulator ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.