Invitrogen
Manufacturer Code:PA5109746
Catalog # PIPA5109746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human FXYD7 (NP_071289.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110035I01Rik FLJ25096 FXYD domain-containing ion transport regulator 7 FXYD7; FXYD domain-containing ion transport regulator 7
Gene Aliases: 1110035I01Rik; FXYD7
UniProt ID: (Human) P58549, (Mouse) P59648, (Rat) P59649
Entrez Gene ID: (Human) 53822, (Mouse) 57780, (Rat) 63848
Molecular Function:
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.