Novus Biologicals
Manufacturer Code:NBP18008220UL
Catalog # NBP18008220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human FXYD7. Peptide sequence MATPTQTPTKAPEEPDPFYYDYNTVQTVGMTLATILFLLGILIVISKKVK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ25096 FXYD domain containing ion transport regulator 7 FXYD domain-containing ion transport regulator 7; FXYD domain-containing ion transport regulator 7
Gene Aliases: FXYD7
UniProt ID: (Human) P58549
Entrez Gene ID: (Human) 53822
Molecular Function:
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.