Invitrogen
Manufacturer Code:PA575640
Catalog # PIPA575640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human FXYD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXTD domain-containing ion transport regulator 2; FXYD domain-containing ion transport regulator 2; gamma subunit of sodium potassium ATPase; Na(+)/K(+) ATPase subunit gamma; Sodium pump gamma chain; sodium/potassium-transporting ATPase gamma chain; Sodium/potassium-transporting ATPase subunit gamma; Sodium/potassium-transporting ATPase subunit gamma Na(+)/K(+) ATPase subunit gamma FXYD domain-containing ion transport regulator 2 Sodium pump gamma chai FXYD2 ATP1C ATP1G1
Gene Aliases: ATP1C; ATP1G1; FXYD2; GNAKATP; HOMG2
UniProt ID: (Human) P54710, (Mouse) Q04646, (Rat) Q04679
Entrez Gene ID: (Human) 486, (Mouse) 11936, (Rat) 29639
Molecular Function:
cation transporter
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.