Invitrogen
Manufacturer Code:PA566670
Catalog # PIPA566670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human FXYD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1C ATP1G1; ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXYD domain-containing ion transport regulator 2; Na(+)/K(+) ATPase subunit gamma; Sodium pump gamma chain; Sodium/potassium-transporting ATPase subunit gamma
Gene Aliases: ATP1C; ATP1G1; FXYD2; HOMG2
UniProt ID: (Human) P54710
Entrez Gene ID: (Human) 486
Molecular Function:
cation transporter
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.