Novus Biologicals
Manufacturer Code:NBP297415AF488
Catalog # NB043112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human FXYD2 fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1C ATP1G1ATPase Na+/K+ transporting gamma 1 polypeptide FXYD domain containing ion transport regulator 2 FXYD domain-containing ion transport regulator 2 HOMG2 hypomagnesemia 2 renal MGC12372 Na(+)/K(+) ATPase subunit gamma Sodium pump gamma chain sodium/potassium-transporting ATPase subunit gamma Sodium-potassium-ATPase gamma polypeptide; ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXYD domain-containing ion transport regulator 2; Na(+)/K(+) ATPase subunit gamma; Sodium pump gamma chain; Sodium/potassium-transporting ATPase subunit gamma
Gene Aliases: ATP1C; ATP1G1; FXYD2; HOMG2
UniProt ID: (Human) P54710
Entrez Gene ID: (Human) 486
Molecular Function: cation transporter enzyme modulator ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.