Invitrogen
Manufacturer Code:PA5109744
Catalog # PIPA5109744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-92 of human FXYD1 (NP_005022.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610012C17Rik 1110006M24Rik FXYD domain-containing ion transport regulator 1 Fxyd1 Phospholemman PLM Pml Sodium Potassium ATPase Sodium/potassium-transporting ATPase subunit FXYD1; FXYD domain-containing ion transport regulator 1; Phospholemman; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Aliases: 0610012C17Rik; 1110006M24Rik; FXYD1; PLM; Pml
UniProt ID: (Human) O00168, (Mouse) Q9Z239, (Rat) O08589
Entrez Gene ID: (Human) 5348, (Mouse) 56188, (Rat) 58971
Molecular Function:
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.