Novus Biologicals
Manufacturer Code:NBP299343H
Catalog # NB084861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FXR1. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; Fragile X mental retardation syndrome-related protein 1; fragile X mental retardation syndrome-related protein 1 fragile X mental retardation autosomal homolog 1 FXR1P hFXR1p; fragile X mental retardation, autosomal homolog 1; hFXR1p
Gene Aliases: FXR1; FXR1P
UniProt ID: (Human) P51114
Entrez Gene ID: (Human) 8087
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.