Novus Biologicals
Manufacturer Code:NB400153AF405
Catalog # NB400153W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt# Q96RI1] |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bile acid receptor; bile acid receptor farnesoid X receptor Farnesoid X-activated receptor Farnesol receptor HRR-1 FXRBAR HRR-1 HRR1MGC163445 Nuclear receptor subfamily 1 group H member 4 nuclear receptor subfamily 1 group H member 4 Retinoid X receptor-interacting protein 14 RIP14RXR-interacting protein 14; farnesoid X nuclear receptor; Farnesoid X-activated receptor; Farnesol receptor HRR-1; Nuclear receptor subfamily 1 group H member 4; Retinoid X receptor-interacting protein 14; RXR-interacting protein 14
Gene Aliases: BAR; FXR; HRR-1; HRR1; NR1H4; RIP14
UniProt ID: (Human) Q96RI1
Entrez Gene ID: (Human) 9971
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.