Invitrogen
Manufacturer Code:PA540755
Catalog # PIPA540755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NR1H4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bile acid receptor; farnesoid X activated receptor; farnesoid X nuclear receptor; Farnesoid X-activated receptor; Farnesol receptor HRR-1; Nuclear receptor subfamily 1 group H member 4; Nuclear receptor subfamily 1 group H member 4 BAR FXR HRR-1 HRR1 MGC163445 RIP14; Retinoid X receptor-interacting protein 14; RXR-interacting protein 14
Gene Aliases: AI957360; BAR; FXR; HRR-1; HRR1; NR1H4; RIP14; Rxrip14
UniProt ID: (Human) Q96RI1, (Bovine) Q3SZL0, (Rat) Q62735, (Mouse) Q60641
Entrez Gene ID: (Horse) 100052821, (Human) 9971, (Bovine) 540528, (Rabbit) 100009485, (Dog) 612928, (Rat) 60351, (Mouse) 20186, (Guinea pig) 100734377
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.