Invitrogen
Manufacturer Code:PA560353
Catalog # PIPA560353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FUT8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 16 Fucosyltransferase alpha1-6FucT FUT8 alpha-(16)-fucosyltransferase alpha1-6FucT EC 2.4.1.68 Fucosyltransferase 8 fucosyltransferase 8 (alpha (16) fucosyltransferase) GDP-fucose--glycoprotein fucosyltransferase GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha16-fucosyltransferase Glycoprotein 6-alpha-L-fucosyltransferase MGC26465; Alpha-(1,6)-fucosyltransferase; Alpha1-6FucT; Fucosyltransferase 8; fucosyltransferase 8 (alpha (1,6) fucosyltransferase); GDP-fucose--glycoprotein fucosyltransferase; GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase; Glycoprotein 6-alpha-L-fucosyltransferase
Gene Aliases: FUT8
UniProt ID: (Human) Q9BYC5
Entrez Gene ID: (Human) 2530
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.