Abnova
Manufacturer Code:H00002529A01
Catalog # 89-104-520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 262 a.a. to 342 a.a) partial recombinant protein with GST tag.,FUT7 (NP_004470 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant FUT7.
For Research Use Only
Protein Aliases: Alpha-(1,3)-fucosyltransferase 7; Fuc-TVII; Fucosyltransferase 7; fucosyltransferase 7 (alpha (1,3) fucosyltransferase); Fucosyltransferase VII; FucT-VII; Galactoside 3-L-fucosyltransferase; Selectin ligand synthase; selectin-ligand synthase
Gene Aliases: FucT-VII; FUT7
UniProt ID: (Human) Q11130
Entrez Gene ID: (Human) 2529
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.