Proteintech
Manufacturer Code:181971AP150UL
Catalog # 50-173-2041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FUT7 Fusion Protein Ag12475 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha (1,3) fucosyltransferase; Alpha (13) fucosyltransferase Fuc TVII Fucosyltransferase 7 Fucosyltransferase VII FucT VII FUT7 Selectin ligand synthase; Alpha-(1,3)-fucosyltransferase 7; Fuc-TVII; Fucosyltransferase 7; fucosyltransferase 7 (alpha (1,3) fucosyltransferase); Fucosyltransferase VII; Galactoside 3-L-fucosyltransferase; Selectin ligand synthase; selectin-ligand synthase
Gene Aliases: AI853193; FTVII; Fuc-TVII; FucT-VII; FUT7
UniProt ID: (Human) Q11130, (Mouse) Q11131, (Rat) Q712G6
Entrez Gene ID: (Human) 2529, (Mouse) 14347, (Rat) 296564
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.