Invitrogen
Manufacturer Code:PA565366
Catalog # PIPA565366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human FUT6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-galactosyl-N-acetylglucosaminide 3-alpha-L-fucosyltransferase FUT6; FCT3A FT1A Fuc-TVI FucT-VI alpha-(1 3)-fucosyltransferase fucosyltransferase VI galactoside 3-L-fucosyltransferase; Fuc-TVI; Fucosyltransferase 6; fucosyltransferase 6 (alpha (1,3) fucosyltransferase); Fucosyltransferase VI; Galactoside 3-L-fucosyltransferase
Gene Aliases: FCT3A; FT1A; Fuc-TVI; FucT-VI; FUT6
UniProt ID: (Human) P51993
Entrez Gene ID: (Human) 2528
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.