Abnova
Manufacturer Code:H00002527M03
Catalog # 89-022-237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H6 |
Immunogen | FUT5 (NP_002025 95 a.a. ~ 164 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant FUT5.
For Research Use Only
Protein Aliases: 3-galactosyl-N-acetylglucosaminide 4-alpha-L-fucosyltransferase FUT5; 4-galactosyl-N-acetylglucosaminide 3-alpha-L-fucosyltransferase FUT5; alpha (1,3) fucosyltransferase; FUC-TV; Fucosyltransferase 5; fucosyltransferase 5 (alpha (1,3) fucosyltransferase); Fucosyltransferase V; fucT-V; Galactoside 3-L-fucosyltransferase
Gene Aliases: FUC-TV; FUT5
UniProt ID: (Human) Q11128
Entrez Gene ID: (Human) 2527
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.