Invitrogen
Manufacturer Code:PA5103133
Catalog # PIPA5103133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FucT-IV corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-fucosyl-N-acetyl-lactosamine epitope; 3-fucosyl-N-acetyl-lactosamine epitope AI451562 alpha (13) fucosyltransferase myeloid specific alpha (13) fucosyltransferase myeloid-specific alpha 13-fucosyltransferase alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4) alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4) alpha-(13)-fucosyltransferase 4 CD15 ELAM ligand fucosyltransferase ELAM-1 ligand fucosyltransferase Elft FAL FCT3A fucosyltransferase 4 fucosyltransferase 4 (alpha (13) fucosyltransferase myeloid-specific) fucosyltransferase IV Fuct Fuc-TIV FucT-IV Fut4 FUTIV Galactoside 3-L-fucosyltransferase Lewis X LeX Ssea1 SSEA-1 stage-specific embryonic antigen 1; alpha (1,3) fucosyltransferase, myeloid specific; alpha (1,3) fucosyltransferase, myeloid-specific; alpha 1,3-fucosyltransferase; alpha 1,3-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4); Alpha-(1,3)-fucosyltransferase 4; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; Fuc-TIV; Fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); Fucosyltransferase IV; fucT-IV; Galactoside 3-L-fucosyltransferase; Lewis X; stage-specific embryonic antigen 1
Gene Aliases: AI451562; CD15; ELFT; FAL; FCT3A; FUC-TIV; Fuct; FucT-IV; FUT4; FUTIV; LeX; SSEA-1; Ssea1
UniProt ID: (Human) P22083, (Mouse) Q11127
Entrez Gene ID: (Human) 2526, (Rat) 60670, (Mouse) 14345
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.