Abnova
Manufacturer Code:H00002524K
Catalog # 89-205-141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human FUT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human FUT2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: alpha (1,2) fucosyltransferase; Alpha(1,2)FT 2; alpha(1,2)FT2; Fucosyltransferase 2; fucosyltransferase 2 (secretor status included); Galactoside alpha-(1,2)-fucosyltransferase 2; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2; Se; SE SEC2 Se2 sej; SE2; Secretor blood group alpha-2-fucosyltransferase; Secretor factor; Type 1 galactoside alpha-(1,2)-fucosyltransferase FUT2; Type 2 galactoside alpha-(1,2)-fucosyltransferase FUT2
Gene Aliases: B12QTL1; FUT2; SE; Se2; SEC2; sej
UniProt ID: (Human) Q10981
Entrez Gene ID: (Human) 2524
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.