Proteintech
Manufacturer Code:171751AP150UL
Catalog # 50-173-1421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FUT11 Fusion Protein Ag10815 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha (1,3) fucosyltransferase; Alpha-(1,3)-fucosyltransferase 11; alpha3-fucosyltransferase 11; Fuc TXI Fucosyltransferase 11 Fucosyltransferase XI FucT XI FUT11; Fuc-TXI; Fucosyltransferase 11; fucosyltransferase 11 (alpha (1,3) fucosyltransferase); Fucosyltransferase XI; fucT-XI; Galactoside 3-L-fucosyltransferase 11
Gene Aliases: 3110009G03Rik; AI317183; FUT11
UniProt ID: (Human) Q495W5, (Mouse) Q8BHC9, (Rat) Q68FV3
Entrez Gene ID: (Human) 170384, (Mouse) 73068, (Rat) 286971
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.