Invitrogen
Manufacturer Code:PA513515
Catalog # PIPA513515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 164-195 amino acids from the central region of human FUT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha(1,2) fucosyltransferase 1; Alpha(1,2)FT 1; Blood group H alpha 2-fucosyltransferase; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); Galactoside 2-alpha-L-fucosyltransferase 1 GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1 Alpha(12)FT 1 Fucosyltransferase 1 Blood group H alpha 2-fucosyltransferase HSC H FUT1; Galactoside alpha-(1,2)-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; Type 1 galactoside alpha-(1,2)-fucosyltransferase FUT1; Type 2 galactoside alpha-(1,2)-fucosyltransferase FUT1
Gene Aliases: FUT1; H; HH; HSC
UniProt ID: (Human) P19526
Entrez Gene ID: (Human) 2523
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.