Invitrogen
Manufacturer Code:PA541929
Catalog # PIPA541929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FUSIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa SR-repressor protein; FUS interacting protein (serine-arginine rich) 1; FUS interacting protein (serine/arginine-rich) 1; Fus-associated protein with serine-arginine repeats; FUS-interacting protein (serine-arginine rich) 2; FUS-interacting serine-arginine-rich protein 1; Neural-salient serine/arginine-rich protein; neural-salient SR protein; Neural-specific SR protein; protein phosphatase 1, regulatory subunit 149; serine-arginine repressor protein (40 kDa); Serine/arginine-rich splicing factor 10; Serine/arginine-rich splicing factor 10 FUSIP2 NSSR SFRS13 SRp38 SRrp40 TASR TASR1 TASR2 FUSIP1 SFRS13A; serine/arginine-rich splicing factor 13A; Splicing factor SRp38; splicing factor, arginine/serine-rich 13; Splicing factor, arginine/serine-rich 13A; SR splicing factor 10; SRrp40; TASR; TLS-associated protein TASR; TLS-associated protein with Ser-Arg repeats; TLS-associated protein with SR repeats; TLS-associated serine-arginine protein; TLS-associated serine-arginine protein 1; TLS-associated serine-arginine protein 2; TLS-associated SR protein
Gene Aliases: FUSIP1; FUSIP2; NSSR; NSSR1; NSSR2; PPP1R149; SFRS13; SFRS13A; SRp38; SRrp40; SRSF10; Srsf13a; TASR; TASR1; TASR2
UniProt ID: (Human) O75494, (Mouse) Q9R0U0
Entrez Gene ID: (Human) 10772, (Horse) 100071526, (Rabbit) 100344333, (Rat) 362630, (Guinea pig) 100721249, (Mouse) 14105
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.