Novus Biologicals
Manufacturer Code:NB11093598SS
Catalog # NB11093598T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of human FUSIP1 (within residues 100-200). [Swiss-Prot# O75494] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa SR-repressor protein; 40 kDa SR-repressor protein FLJ43846 FUS interacting protein (serine-arginine rich) 1 FUS-interacting protein (serine-arginine rich) 2 FUS-interacting serine-arginine-rich protein 1 FUSIP1DKFZp686H0644 FUSIP2 neural-salient SR protein NSSR serine/arginine-rich splicing factor 10 serine-arginine repressor protein (40 kDa) SFRS13 SFRS13A Splicing factor SRp38 splicing factor arginine/serine-rich 13 Splicing factor arginine/serine-rich 13ATLS-associated protein with Ser-Arg repeats SR splicing factor 10 SRp38 SRrp40TLS-associated protein with SR repeats TASR1TLS-associated SR protein TASR2FUS interacting protein (serine/arginine-rich) 1 TASRFLJ30749 TLS-associated protein TASR TLS-associated serine-arginine protein TLS-associated serine-arginine protein 1 TLS-associated serine-arginine protein 2; FUS interacting protein (serine-arginine rich) 1; FUS-interacting protein (serine-arginine rich) 2; FUS-interacting serine-arginine-rich protein 1; neural-salient SR protein; protein phosphatase 1, regulatory subunit 149; serine-arginine repressor protein (40 kDa); Serine/arginine-rich splicing factor 10; Splicing factor SRp38; splicing factor, arginine/serine-rich 13; Splicing factor, arginine/serine-rich 13A; SR splicing factor 10; SRrp40; TASR; TLS-associated protein TASR; TLS-associated protein with Ser-Arg repeats; TLS-associated protein with SR repeats; TLS-associated serine-arginine protein; TLS-associated serine-arginine protein 1; TLS-associated serine-arginine protein 2; TLS-associated SR protein
Gene Aliases: FUSIP1; FUSIP2; NSSR; PPP1R149; SFRS13; SFRS13A; SRp38; SRrp40; SRSF10; TASR; TASR1; TASR2
UniProt ID: (Human) O75494
Entrez Gene ID: (Human) 10772
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.