Novus Biologicals
Manufacturer Code:NBP141157AF750
Catalog # NP14117A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F4 |
Immunogen | Recombinant FUS2 (1-308aa) purified from E. coli |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FUS-2 FUS2EC 2.3.1.- N-acetyltransferase 6 N-acetyltransferase 6 (GCN5-related) Protein fus-2 Protein fusion-2; HsNAAA80; N-acetyltransferase 6; N-acetyltransferase 6 (GCN5-related); N-alpha-acetyltransferase 80; Protein fus-2; Protein fusion-2
Gene Aliases: FUS-2; FUS2; NAA80; NAT6
UniProt ID: (Human) Q93015
Entrez Gene ID: (Human) 24142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.