Invitrogen
Manufacturer Code:PA558844
Catalog # PIPA558844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FUS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FUS-2 FUS2 N-acetyltransferase 6 protein fusion-2; fusion 2; HsNAAA80; N-acetyltransferase 6; N-acetyltransferase 6 (GCN5-related); N-acetyltransferase 6-like; N-alpha-acetyltransferase 80; Protein fus-2; Protein fusion-2; Protein fusion-2 homolog
Gene Aliases: AI225910; FUS-2; FUS2; NAA80; NAT6
UniProt ID: (Human) Q93015, (Mouse) Q9R123
Entrez Gene ID: (Rat) 100910872, (Human) 24142, (Mouse) 56441
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.