Novus Biologicals
Manufacturer Code:NB100562
Catalog # NB100562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide which maps to a region between residues 500 and the C-terminus (residue 526) of human fusion (involved in t(1216) in malignant liposarcoma) using the numbering given in SwissProt entry P35637 (GeneID 2521). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75 kDa DNA-pairing protein; 75 kDa DNA-pairing protein ALS6 amyotrophic lateral sclerosis 6 FUS1 fused in sarcoma fusion (involved in t(1216) in malignant liposarcoma) fusion gene in myxoid liposarcoma fusion derived from t(1216) malignant liposarcoma fus-like protein heterogeneous nuclear ribonucleoprotein P2 HNRNPP2 Oncogene FUS Oncogene TLS POMp75 RNA-binding protein FUS TLShnRNP-P2 Translocated in liposarcoma protein; fus-like protein; fused in sarcoma; fusion gene in myxoid liposarcoma; heterogeneous nuclear ribonucleoprotein P2; Oncogene FUS; Oncogene TLS; POMp75; RNA-binding protein FUS; Translocated in liposarcoma protein
Gene Aliases: ALS6; ETM4; FUS; FUS1; HNRNPP2; POMP75; TLS
UniProt ID: (Human) P35637
Entrez Gene ID: (Human) 2521
Molecular Function:
DNA binding protein
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.