Invitrogen
Manufacturer Code:PA572155
Catalog # PIPA572155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 122-150 amino acids from the C-terminal region of human FUNDC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FUN14 domain-containing protein 2; FUN14 domain-containing protein 2 Cervical cancer proto-oncogene 3 protein HCC-3 Hepatitis C virus core-binding protein 6 FUNDC2 HCBP6; Hepatitis C virus core-binding protein 6
Gene Aliases: 4833415N24Rik; DC44; Fundc2; HCBP6
UniProt ID: (Mouse) Q9D6K8
Entrez Gene ID: (Mouse) 67391
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.