Invitrogen
Manufacturer Code:PA577785
Catalog # PIPA577785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the N-terminal of Human FUNDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FUN14 domain-containing protein 1; FUNDC1 FUN14 domain-containing protein 1
Gene Aliases: 1500005J14Rik; 1810033P05Rik; FUNDC1
UniProt ID: (Human) Q8IVP5, (Mouse) Q9DB70, (Rat) Q5BJS4
Entrez Gene ID: (Human) 139341, (Mouse) 72018, (Rat) 363442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.