Proteintech
Manufacturer Code:164201AP150UL
Catalog # 50-173-0933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FUCA1 Fusion Protein Ag9612 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha L fucosidase 1 Alpha L fucosidase I FUCA FUCA1 fucosidase alpha L 1 tissue Tissue alpha L fucosidase; Alpha-L-fucosidase 1; Alpha-L-fucosidase I; Alpha-L-fucoside fucohydrolase 1; Fucosidase alpha-L-1 tissue; Fucosidase, alpha-L-1, tissue; Tissue alpha-L-fucosidase
Gene Aliases: 0610006A03Rik; 9530055J05Rik; Afuc; FUCA; FUCA1; Nbla10230
UniProt ID: (Human) P04066, (Rat) P17164, (Mouse) Q99LJ1
Entrez Gene ID: (Human) 2517, (Rat) 24375, (Mouse) 71665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.