Invitrogen
Manufacturer Code:PA544281
Catalog # PIPA544281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FTSJD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cap methyltransferase 1; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; FtsJ methyltransferase domain containing 2; FtsJ methyltransferase domain containing 2 KIAA0082 MTr1 hMTr1 FTSJD2; FtsJ methyltransferase domain-containing protein 2; Interferon-stimulated gene 95 kDa protein; ISG95; MTr1; S-adenosyl-L-methionine-dependent methyltransferase FTSJD2
Gene Aliases: 1300018I05Rik; AI451264; CMTR1; FTSJD2; hMTr1; KIAA0082; LRRG00129; mKIAA0082; MTR1; RGD1307801; zgc:55336
UniProt ID: (Human) Q8N1G2, (Bovine) A2VE39, (Rat) Q5U2Z5, (Mouse) Q9DBC3, (Zebrafish) Q803R5
Entrez Gene ID: (Rabbit) 100350011, (Pig) 100154061, (Dog) 100683999, (Human) 23070, (Bovine) 509620, (Horse) 100064496, (Rat) 309656, (Guinea pig) 100725670, (Mouse) 74157, (Zebrafish) 393102
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.