Novus Biologicals
Manufacturer Code:NBP183047
Catalog # NBP183047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SQGRKDIVEASSQKGRRGLGLTLRGFDQELNVDWRDEPEPSACEQVSWFPECTTEIPDTQEMSDWMVVGKRKMIIEDETEFCGEELLHSVLQCK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cap methyltransferase 1; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; Cap1 2'O-ribose methyltransferase 1 cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1 EC 2.1.1.57 FtsJ methyltransferase domain containing 2 FtsJ methyltransferase domain-containing protein 2 hMTr1 Interferon-stimulated gene 95 kDa protein ISG95 KIAA0082FLJ22156 MTR1 S-adenosyl-L-methionine-dependent methyltransferase FTSJD2; FtsJ methyltransferase domain containing 2; FtsJ methyltransferase domain-containing protein 2; Interferon-stimulated gene 95 kDa protein; ISG95; MTr1; S-adenosyl-L-methionine-dependent methyltransferase FTSJD2
Gene Aliases: CMTR1; FTSJD2; hMTr1; KIAA0082; MTR1
UniProt ID: (Human) Q8N1G2
Entrez Gene ID: (Human) 23070
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.