Invitrogen
Manufacturer Code:PA570295
Catalog # PIPA570295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FTSJD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrift homolog; Cap methyltransferase 2; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 2; Cap2 2'O-ribose methyltransferase 2; FtsJ methyltransferase domain containing 1; FtsJ methyltransferase domain-containing protein 1; HMTr2; MTr2; Protein adrift homolog
Gene Aliases: AFT; AU022703; C730036L12Rik; CMTR2; FTSJD1; HMTr2; MTr2; RGD1309394
UniProt ID: (Human) Q8IYT2, (Mouse) Q8BWQ4
Entrez Gene ID: (Human) 55783, (Horse) 100068485, (Rabbit) 100346528, (Dog) 489718, (Rat) 292016, (Mouse) 234728
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.