Novus Biologicals
Manufacturer Code:NBP230944
Catalog # NBP230944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: DKVAKGYFNSWAEEHGVYHPGQSSILEGTASNLECHLWHILEGKKLPKVKCSPFCNGEILKTLNEAIEKSLGGAFNLDSKFRPKQQYSCSCH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrift homolog; adrift homolog AFTftsJ methyltransferase domain-containing protein 1 EC 2.1.1 EC 2.1.1.- FLJ11171 FtsJ methyltransferase domain containing 1 Protein adrift homolog; Cap methyltransferase 2; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 2; Cap2 2'O-ribose methyltransferase 2; FtsJ methyltransferase domain containing 1; FtsJ methyltransferase domain-containing protein 1; HMTr2; Protein adrift homolog
Gene Aliases: AFT; CMTR2; FTSJD1; HMTr2; MTr2
UniProt ID: (Human) Q8IYT2
Entrez Gene ID: (Human) 55783
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.