Invitrogen
Manufacturer Code:PA554090
Catalog # PIPA554090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FTSJ2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16S rRNA (uridine(1369)-2'-O)-methyltransferase; 16S rRNA [Um1369] 2'-O-methyltransferase; cell division protein FtsJ; epididymis luminal protein 97; FJH1 cell division protein FtsJ protein ftsJ homolog 2 putative ribosomal RNA methyltransferase 2 rRNA (uridine-2&prime-O-)-methyltransferase; FtsJ homolog 2; FtsJ RNA methyltransferase homolog 2; MRM2 RNA methyltransferase homolog; Protein ftsJ homolog 2; putative ribosomal RNA methyltransferase 2; rRNA (uridine-2'-O-)-methyltransferase; rRNA methyltransferase 2, mitochondrial
Gene Aliases: FJH1; FTSJ2; HEL97; MRM2; RRMJ2
UniProt ID: (Human) Q9UI43
Entrez Gene ID: (Human) 29960
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.