Abnova
Manufacturer Code:H00029960B01P
Catalog # 89-934-169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | FTSJ2 (NP_037525.11 a.a. &bsim 246 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human FTSJ2 protein.
For Research Use Only
Protein Aliases: 16S rRNA (uridine(1369)-2'-O)-methyltransferase; 16S rRNA [Um1369] 2'-O-methyltransferase; cell division protein FtsJ; DKFZp686J14194; epididymis luminal protein 97; FJH1; FtsJ homolog 2; FtsJ RNA methyltransferase homolog 2; MRM2 RNA methyltransferase homolog; Protein ftsJ homolog 2; putative ribosomal RNA methyltransferase 2; rRNA (uridine-2'-O-)-methyltransferase; rRNA methyltransferase 2, mitochondrial
Gene Aliases: FJH1; FTSJ2; HEL97; MRM2; RRMJ2
UniProt ID: (Human) Q9UI43
Entrez Gene ID: (Human) 29960
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.