Proteintech
Manufacturer Code:225561AP150UL
Catalog # 50-173-3888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FTSJ2 Fusion Protein Ag18410 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16S rRNA (uridine(1369)-2'-O)-methyltransferase; 16S rRNA [Um1369] 2'-O-methyltransferase; cell division protein FtsJ; epididymis luminal protein 97; FJH1 FtsJ homolog 2 (E. coli) FTSJ2 Protein ftsJ homolog 2; FtsJ homolog 2; FtsJ RNA methyltransferase homolog 2; MRM2 RNA methyltransferase homolog; Protein ftsJ homolog 2; putative ribosomal RNA methyltransferase 2; rRNA (uridine-2'-O-)-methyltransferase; rRNA methyltransferase 2, mitochondrial
Gene Aliases: 2310037B18Rik; FJH1; FTSJ2; HEL97; MRM2; RRMJ2
UniProt ID: (Mouse) Q9CPY0, (Human) Q9UI43
Entrez Gene ID: (Mouse) 68017, (Human) 29960
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.