Invitrogen
Manufacturer Code:PA518942
Catalog # PIPA518942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QQKPDCRPYWEKDD) corresponding to the internal amino acids of Fto |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AlkB homolog 9; Alpha-ketoglutarate-dependent dioxygenase FTO; Fat mass and obesity-associated protein; Fto fat mass and obesity associated AW743446 mKIAA1752 similar to FTO; m6A(m)-demethylase FTO; mRNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; mRNA N(6)-methyladenosine demethylase FTO; tRNA N1-methyl adenine demethylase FTO; U6 small nuclear RNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; U6 small nuclear RNA N(6)-methyladenosine-demethylase FTO
Gene Aliases: ALKBH9; BMIQ14; FTO; GDFD; KIAA1752
UniProt ID: (Human) Q9C0B1
Entrez Gene ID: (Human) 79068
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.