Invitrogen
Manufacturer Code:PA530906
Catalog # PIPA530906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 37 and 242 of Human FTMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ferritin H; ferritin H subunit; ferritin heavy chain 3; Ferritin, mitochondrial; mitochondrial ferritin; MTF ferritin H subunit ferritin mitochondrial mitochondrial ferritin
Gene Aliases: 4930447C24Rik; FerH; Fth3; FTMT; MTF
UniProt ID: (Human) Q8N4E7, (Mouse) Q9D5H4
Entrez Gene ID: (Human) 94033, (Mouse) 67634
Molecular Function: storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.